Få mere at vide
Droperidol EP Impurity C (as Chloride), Mikromol™
Artikelnummer. 30051923
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30051923 25 mg
1 options
Artikelnummer. 30051923 Leverandør Mikromol Leverandørnr. MM0138.030025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Droperidol EP Impurity C (as Chloride)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-[4-(4-Fluorophenyl)-4-oxo-butyl]-4-(2-oxo-2,3-dihydro-1H-benzimidazol-1-yl)pyridinium Chloride
CAS 60373-76-4
Analyt- eller komponentnavne 1-[4-(4-Fluorophenyl)-4-oxo-butyl]-4-(2-oxo-2,3-dihydro-1H-benzimidazol-1-yl)pyridinium Chloride
Forsendelsestilstand Room Temperature
Molekylær formel C22 H19 F N3 O2 . Cl
Certificeringer/compliance ISO 17025
Synonym Droperidol Imp. C (EP), 1-[4-(4-Fluorophenyl)-4-oxo-butyl]-4-(2-oxo-2,3-dihydro-1H-benzimidazol-1-yl)pyridinium Chloride, 4-(2,3-Dihydro-2-oxo-1H-benzimidazol-1-yl)-1-[4-(4-fluorophenyl)-4-oxobutyl]-pyridinium chloride, Droperidol Impurity C as Chloride
InChI formel InChI=1S/C22H18FN3O2.ClH/c23-17-9-7-16(8-10-17)21(27)6-3-13-25-14-11-18(12-15-25)26-20-5-2-1-4-19(20)24-22(26)28;/h1-2,4-5,7-12,14-15H,3,6,13H2;1H
SMIL [Cl-].Fc1ccc(cc1)C(=O)CCC[n+]2ccc(cc2)N3C(=O)Nc4ccccc34
IUPAC navn 3-[1-[4-(4-fluorophenyl)-4-oxobutyl]pyridin-1-ium-4-yl]-1H-benzimidazol-2-one;chloride
Molekylvægt (g/mol) 411.86
Mængde 25 mg
Formel vægt 411.115 g/mol