Få mere at vide
Droloxifene Citrate, TRC
Artikelnummer. 30138052
Skift visning
Mængde:
10 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138052 100 mg
30135246 500 mg
30005518 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30138052 Leverandør Toronto Research Chemicals Leverandørnr. TRCD679200100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 97752-20-0
Molekylær formel C26 H29 N O2 . C6 H8 O7
Molekylvægt (g/mol) 579.64
Synonym 3-[(1E)-1-[4-[2-(Dimethylamino)ethoxy]phenyl]-2-phenyl-1-buten-1-yl]phenol 2-Hydroxy-1,2,3-propanetricarboxylate (1:1);
IUPAC navn 3-[(Z)-1-[4-[2-(dimethylamino)ethoxy]phenyl]-2-phenylbut-1-enyl]phenol;2-hydroxypropane-1,2,3-tricarboxylic acid
SMIL CC\C(=C(/c1ccc(OCCN(C)C)cc1)\c2cccc(O)c2)\c3ccccc3.OC(=O)CC(O)(CC(=O)O)C(=O)O
Tekniske data
Kemisk navn eller materiale Droloxifene Citrate
Mængde 100 mg
InChI formel InChI=1S/C26H29NO2.C6H8O7/c1-4-25(20-9-6-5-7-10-20)26(22-11-8-12-23(28)19-22)21-13-15-24(16-14-21)29-18-17-27(2)3;7-3(8)1-6(13,5(11)12)2-4(9)10/h5-16,19,28H,4,17-18H2,1-3H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)/b26-25-;
Formel vægt 579.2468