Learn More
Dräger™ Short-Term Detector Tubes - Phosgene
1640.00 DKK - 1655.00 DKK
Product Code | Brand | Components Measured | Measurement Range | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Components Measured | Measurement Range | Price | Quantity & Availability | ||||
13197276
|
Dräger™
CH28301 |
Phosgene 0.25/b | 0.25 to 5ppm |
1655.00 DKK
Pack of 10 |
Estimated Shipment: 30-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11368291
|
Dräger™
8101521 |
Phosgene 0.02/a | 0.02 to 1ppm |
1640.00 DKK
Each |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
1Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
2Safety Equipment Institute (SEI) certified.
Specifications
3 to 15mg/L | |
10 |