Learn More
Dräger™ CMS Chips: Methanol
Suitable for 10 measurements of one gas
Brand: Dräger™ 6406380
Additional Details : Weight : 0.02880kg
Description
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.Specifications
700 to 1100 hpa | |
5 to 25mg/L | |
200 to 600 sec. | |
15°C to 30°C |
Methanol CMS Chip | |
20 to 500 | |
59°F to 104°F |