missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
DPPIV/CD26 Antibody (236.3), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22434MFV450
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
DPPIV/CD26 Monoclonal antibody specifically detects DPPIV/CD26 in Human, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Tekniske data
| DPPIV/CD26 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation | |
| 236.3 | |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation | |
| ADABP, ADCP-2, ADCP2DPP IV, Adenosine deaminase complexing protein 2TP103, CD26 antigen, CD26T-cell activation antigen CD26, dipeptidyl peptidase 4, Dipeptidyl peptidase IV, dipeptidylpeptidase 4, dipeptidyl-peptidase 4, dipeptidylpeptidase IV (CD26, adenosine deaminase complexing protein 2), DPPIV, EC 3.4.14.5 | |
| Rat 110-120 kD single chain glycoprotein Depeptidyl Peptidase IV (CD26). | |
| 0.1 mL | |
| Adaptive Immunity, Cellular Markers, GPCR, Immunology | |
| 1803 | |
| Store at 4C in the dark. | |
| IgG2b κ |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion