Få mere at vide
Tocris Bioscience™ DPN
GREENER_CHOICE
Artikelnummer. 30001261
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30001261 10 mg
18055052 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30001261 Leverandør Tocris Bioscience™ Leverandørnr. 1494/10
Highly potent ERβ agonist
Highly potent estrogen ERβ receptor agonist with a 70-fold selectivity over ERα (EC50 values are 0.85 and 66 nM respectively). Relaxes mesenteric arteries in vitro. Shown to regulate expression of GluR1, GluR2 and GluR3 in rat hippocampus.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1428-67-7
Molekylær formel C15H13NO2
Tekniske data
Kemisk navn eller materiale DPN
Mængde 10 mg
Formel vægt 239.27
Renhed >99%
Molekylvægt (g/mol) 239.274