Få mere at vide
DPF3 Antibody [mFluor Violet 500 SE], Novus Biologicals Biologicals™
Handla allt Bio Techne ProdukterBeskrivelse
DPF3 Polyclonal antibody specifically detects DPF3 in Mouse,Rat samples. It is validated for ELISA,Western Blot
Tekniske data
Tekniske data
| Antigen | DPF3 |
| Applikationer | ELISA, Western Blot |
| Klassifikation | Polyclonal |
| Konjugeret | mFluor Violet 500 SE |
| Formulering | 50mM Sodium Borate |
| Gene Alias | CERD4, D4, zinc and double PHD fingers, family 3 |
| Værtsarter | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 90-200 of human DPF3 (NP_036206.3).,, Sequence:, DPKLRLLEIKPEVELPLKKDGFTSESTTLEALLRGEGVEKKVDAREEESIQEIQRVLENDENVEEGNEEEDLEEDIPKRKNRTRGRARGSAGGRRRHDAASQEDHDKPYVC |
| Oprensningsmetode | Affinity purified |
| Mængde | 0.1 mL |
| Vis mere |