Få mere at vide
DPBQ, MedChemExpress
Artikelnummer. 16462648
Encompass_Preferred
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
16462648 5 mg
1 options
Artikelnummer. 16462648 Leverandør Medchem Express Leverandørnr. HYU00441
MedChemExpress DPBQ activates p53 and triggers apoptosis in a polyploid-specific manner, but does not inhibit topoisomerase or bind DNA. DPBQ elicits expression and phosphorylation of p53 and this effect is specific to tetraploid cells.
DPBQ activates p53 and triggers apoptosis in a polyploid-specific manner, but does not inhibit topoisomerase or bind DNA. DPBQ elicits expression and phosphorylation of p53 and this effect is specific to tetraploid cells.
Tekniske data
Kemisk navn eller materiale DPBQ
CAS 7029-89-2
Farve Green
Renhedsgrad noter Research
Molekylær formel C24H14N2O2
Mængde 5 mg
Opløselighedsinformation DMSO : 2.94 mg/mL (8.11 mM; ultrasonic and warming and heat to 80°C)
SMIL O=C(C1=CC=CC=C12)C3=C(N=C(C4=CC=CC=C4)C(C5=CC=CC=C5)=N3)C2=O
Molekylvægt (g/mol) 362.38
Formel vægt 362.38
Procent renhed 98.0%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-Kinase/protease