Få mere at vide
Doxofylline-d6 (dimethyl-d6), CDN
Artikelnummer. 30319901
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30319901 0.01 g
30311218 0.005 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319901 Leverandør CDN Leverandørnr. CDND71740.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Doxofylline-D6 (dimethyl-D6)
CAS 1219805-99-8
Molekylær formel C11 D6 H8 N4 O4
Mængde 0.01 g
Synonym Doxofylline-d6 (dimethyl-d6), 7-(1,3-Dioxolan-2-ylmethyl)-3,7-dihydro-1,3-di(methyl-d3)-1 H-purine-2,6-dione, 7-(1,3-Dioxolan-2-ylmethyl)theophylline-d6
InChI formel InChI=1 S/C11H14N4O4/c1-13-9-8(10(16)14(2)11(13)17)15(6-12-9)5-7-18-3-4-19-7/h6-7 H,3-5H2,1-2H3/i1D3,2D3
SMIL [2 H]C([2 H])([2 H])N1C(=O)N(c2ncn(CC3OCCO3)c2C1=O)C([2 H])([2 H])[2 H]
IUPAC navn 7-(1,3-dioxolan-2-ylmethyl)-1,3-bis(trideuteriomethyl)purine-2,6-dione
Molekylvægt (g/mol) 272.2902
Formel vægt 272.1392 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity