Få mere at vide
Doxercalciferol, MedChemExpress
Artikelnummer. 16497806
Encompass_Preferred
Skift visning
Mængde:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16497806 100 mg
16480336 2 mg
16463957 50 mg
16460498 5 mg
16448038 10 mg
16443746 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Artikelnummer. 16497806 Leverandør Medchem Express Leverandørnr. HY32348
MedChemExpress Doxercalciferol is a Vitamin D2 analog, acts as an activator of Vitamin D receptor, and prevent renal disease.
Doxercalciferol is a Vitamin D2 analog, acts as an activator of Vitamin D receptor, and prevent renal disease.
Tekniske data
Kemisk navn eller materiale Doxercalciferol
CAS 54573-75-0
Farve White
Renhedsgrad noter Research
Molekylær formel C28H44O2
Mængde 100 mg
Synonym 1.alpha.-Hydroxyvitamin D2
Opløselighedsinformation DMSO : ≥ 50 mg/mL (121.17 mM)
SMIL C=C1[C@H](C[C@@H](C/C1=C/C=C2[C@]3([C@@](C)([C@H](CC3)[C@@H](/C=C/[C@@H](C(C)C)C)C)CCC/2)[H])O)O
Molekylvægt (g/mol) 412.65
Formel vægt 412.65
Procent renhed 95.0%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-programmed cell death