Få mere at vide
(±)-Doxazosin-d8 (piperazine-d8), CDN
Artikelnummer. 30319221
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30319221 0.005 g
30309809 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319221 Leverandør CDN Leverandørnr. CDND60150.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Doxazosin-D8 (piperazine-D8)
CAS 1126848-44-9
Molekylær formel C23 D8 H17 N5 O5
Mængde 0.005 g
Synonym Methanone, [4-(4-amino-6,7-dimethoxy-2-quinazolinyl)-1-piperazinyl-2,2,3,3,5,5,6,6-d8](2,3-dihydro-1,4-benzodioxin-2-yl)- (ACI), [4-(4-Amino-6,7-dimethoxy-2-quinazolinyl)-1-piperazinyl-2,2,3,3,5,5,6,6-d8](2,3-dihydro-1,4-benzodioxin-2-yl)methanone (ACI), Doxazosin-d8 (piperazine-d8), Doxazosin-d8, Doxazosin-D8 (piperazine-D8), 1-(4-Amino-6,7-dimethoxy-2-quinazolinyl)-4-[(2,3-dihydro-1,4-benzodioxin-2-yl)carbonyl]piperazine-2,2,3,3,5,5,6,6-d8, 4-Amino-2-[4-(1,4-benzodioxan-2-carbonyl)-2,2,3,3,5,5,6,6-octadeuteriopiperazin-1-yl]-6,7-dimethoxyquinazoline, [4-(4-Amino-6,7-dimethoxy-2-quinazolinyl)-1-piperazinyl-d8](2,3-dihydro-1,4-benzodioxin-2-yl)methanone
InChI formel InChI=1 S/C23H25N5O5/c1-30-18-11-14-15(12-19(18)31-2)25-23(26-21(14)24)28-9-7-27(8-10-28)22(29)20-13-32-16-5-3-4-6-17(16)33-20/h3-6,11-12,20 H,7-10,13H2,1-2H3,(H2,24,25,26)/i7D2,8D2,9D2,10D2
SMIL [2 H]C1([2 H])N(C(=O)C2COc3ccccc3O2)C([2 H])([2 H])C([2 H])([2 H])N(c4nc(N)c5cc(OC)c(OC)cc5n4)C1([2 H])[2 H]
IUPAC navn [4-(4-amino-6,7-dimethoxyquinazolin-2-yl)-2,2,3,3,5,5,6,6-octadeuteriopiperazin-1-yl]-(2,3-dihydro-1,4-benzodioxin-3-yl)methanone
Molekylvægt (g/mol) 459.5244
Formel vægt 459.2358 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity