Learn More
Cole-Parmer™ Double open end silver/carbon cartridge
Remove solids, organic compounds, chlorine, tastes, and odors with this cartridge. Recommended as pretreatment to DI systems, cycling systems with long down times and other applications where effective media carbon filtration is required.
Brand: Cole-Parmer™ 01508-91
Additional Details : Weight : 0.80000kg
Description
- Silver/Carbon Cartridge with double open end
- Silver impregnated in activated carbon to inhibit growth of bacteria
- For cold water only
- Produces 3500 gallons prior to exhaustion
- Max temperature is 77°F (25°C)
- Max differential pressure is 10psid
- Note: Cartridge does not work with DOE housing models 01508-40, -41, and -42.
Specifications
1 GPM | |
Activated Carbon | |
Double Open End Silver/Carbon Cartridge | |
25.4 cm |
25 μm | |
32°C | |
10 in. |