Learn More
Bochem™ Double Ended Micro Spatula
Bochem™ Double Ended Micro Spatula for handling and sampling small samples.
Brand:  Bochem™ 3012
Additional Details : Weight : 0.09000kg
 View more versions of this product
Product Code. 10060831
  • 17.25 DKK / Each
Estimated Shipment: 05-06-2024
to see stock.
Add to basket
Description
Description
  • Double ended
  • Stainless Steel
Specifications
Specifications
Double Ended
40 mm
Stainless Steel (18/10)
Micro Spatula
2 mm
150 mm