Få mere at vide
Dothiepin-D3, TRC
Artikelnummer. 30373348
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30373348 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30373348 Leverandør Toronto Research Chemicals Leverandørnr. TRCD5365021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Dothiepin-D3
CAS 136765-31-6
Molekylær formel C19H18D3NS
Mængde 1 mg
Synonym 1-Propanamine, 3-dibenzo[b,e]thiepin-11(6H)-ylidene-N-methyl-N-(methyl-d3)- (9CI), Dibenzo[b,e]thiepin, 1-propanamine deriv. (ZCI), 3-Dibenzo[b,e]thiepin-11(6H)-ylidene-N-methyl-N-(methyl-d3)-1-propanamine, Dothiepin-d3, Dosulepin-d3, Dothiepin-d3 (N-methyl-d3) (cis/trans mixture), (EZ)-Dosulepin-D3 (N-methyl-D3)
InChI formel InChI=1S/C19H21NS/c1-20(2)13-7-11-17-16-9-4-3-8-15(16)14-21-19-12-6-5-10-18(17)19/h3-6,8-12H,7,13-14H2,1-2H3/b17-11+/i5D,6D,10D
SMIL [2H]c1cc2SCc3ccccc3\C(=C/CCN(C)C)\c2c([2H])c1[2H]
Molekylvægt (g/mol) 298.46