Få mere at vide
Dosulepin EP Impurity A, Mikromol™
Artikelnummer. 30050642
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30050642 25 mg
17973326 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30050642 Leverandør Mikromol Leverandørnr. MM0510.010025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Dosulepin EP Impurity A
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Dosulepin Sulfoxide
CAS 42046-35-5
Analyt- eller komponentnavne Dosulepin Sulfoxide
Forsendelsestilstand Room Temperature
Molekylær formel C19 H21 N O S
Certificeringer/compliance ISO 17025
Synonym (E)-3-(5-Oxo-5λ4-dibenzo[b,e]thiepin-11(6H)-ylidene)-N,N-dimethylpropan-1-amine (Dosulepin Sulfoxide), 1-Propanamine, N,N-dimethyl-3-(5-oxidodibenzo[b,e]thiepin-11(6H)-ylidene)-, (E)- (9CI), 1-Propanamine, 3-dibenzo[b,e]thiepin-11(6H)-ylidene-N,N-dimethyl-, S-oxide, (E)-, Dibenzo[b,e]thiepin, 1-propanamine deriv., Dosulepin Sulfoxide, Dosulepin Imp. A (EP), (E)-3-(6,11-dihydrodibenzo[b,e]thiepin-11-ylidene)-N,N'- dimethylpropan-1-amine S-oxide, (E)-3-(5-Oxo-5lambda4-dibenzo[b,e]thiepin-11(6H)-ylidene)-N,N-dimethylpropan-1-amine, Dosulepin Hydrochloride Impurity A, Dosulepin Impurity A
InChI formel InChI=1S/C19H21NOS/c1-20(2)13-7-11-17-16-9-4-3-8-15(16)14-22(21)19-12-6-5-10-18(17)19/h3-6,8-12H,7,13-14H2,1-2H3/b17-11+
SMIL CN(C)CC\C=C\1/c2ccccc2CS(=O)c3ccccc13
IUPAC navn Dosulepin Sulfoxide
Molekylvægt (g/mol) 311.44
Mængde 25 mg
Formel vægt 311.1344 g/mol