Learn More
Miele Professional™ Dosing System
Hose dispenser pump, with level indicator for the automatic dispensing of liquid cleaning agents
4910.00 DKK
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
15487714
|
Miele Professional™
9968660 |
200 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15477714
|
Miele Professional™
9964530 |
333 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- For Miele glass laboratory washer PG 8504
- Dispenser module for liquid detergent
- 20cm siphon for canister
- 'Container empty' indicator
Specifications
Gray | |
200/230 V |