missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Donkey F(ab')2 anti-Human IgG (H+L) Secondary Antibody (Pre-adsorbed), Novus Biologicals™
Donkey Polyclonal Antibody
Brand: Novus Biologicals NBP2-62584
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Tekniske data
| IgG (H+L) | |
| Polyclonal | |
| Western Blot 1:2000-1:10000, ELISA 1:20000, Immunohistochemistry 1:1000 - 1:5000 | |
| Donkey | |
| Multi-step | |
| RUO | |
| Secondary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 | |
| Human IgG whole molecule | |
| 0.5 mg | |
| Adaptive Immunity, Immunology | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion