missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Donkey anti-Rat IgG (H+L) Secondary Antibody, Biotin, Novus Biologicals™
Donkey Polyclonal Antibody
Brand: Novus Biologicals NBP1-73148-2000UG
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Spezifikation
| IgG (H+L) | |
| Polyclonal | |
| Biotin | |
| Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free | |
| Rat IgG whole molecule | |
| 2000 μg | |
| Reconstitute with 1.0 mL deionized water | |
| Store at 4°C in the dark. |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| LYOPH | |
| Western Blot 1:2000 - 1:10000, ELISA 1:20000 - 1:100000, Immunohistochemistry 1:1000 - 1:5000, Immunohistochemistry-Paraffin 1:1000 - 1:5000, Immunohistochemistry-Frozen 1:1000 - 1:5000 | |
| Donkey | |
| Affinity Purified | |
| Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-biotin, anti-Donkey Serum, Rat IgG and Rat Serum. | |
| Rat | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur