missing translation for 'onlineSavingsMsg'
Få mere at vide

Donkey anti-Rabbit IgG (H+L) Secondary Antibody, DyLight 650 (Pre-adsorbed), Novus Biologicals™

Artikelnummer. 18287575 Shop alle Bio Techne produkter
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18287575 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18287575 Leverandør Novus Biologicals Leverandørnr. NBP175644

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Donkey Polyclonal Antibody

IgG (H+L) Polyclonal specifically detects IgG (H+L) in Rabbit samples. It is validated for Flow Cytometry, Immunomicroscopy.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen IgG (H+L)
Applikationer Flow Cytometry, Immunomicroscopy
Klassifikation Polyclonal
Koncentration LYOPH
Konjugeret DyLight 650
Fortynding Flow Cytometry 1:20 - 1:2000, Immunomicroscopy 1:20 - 1:2000
Værtsarter Donkey
Immunogen Purified Rabbit IgG (H&L)
Oprensningsmetode Affinity purified
Mængde 1 mg
Regulatorisk status RUO
Forskningsdisciplin Adaptive Immunity, Immunology
Primær eller sekundær Secondary
Testspecificitet Based on IEP, this antibody reacts with heavy gamma chains on rabbit IgG and light chains on all rabbit immunoglobulins. This antibody has been pre-adsorbed against bovine, chicken, goat, guinea pig, hamster, horse, human, mouse, rat and sheep IgG.
Rekonstitution Rehydrate with 1.1 ml of deionized water and let stand 30 minutes at room temperature to dissolve. (Product has been overfilled to ensure complete recovery.) Centrifuge to remove any particulates. Prepare fresh working dilution daily.
Målarter Rabbit
Indhold og opbevaring Store lyophilized antibody at 4°C. Mix reconstituted liquid with an equal volume of glycerol, aliquot contents and freeze at -20°C or below for long term storage.
Isotype IgG
Vis mere Vis mindre

For Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.