missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Donkey anti-Mouse IgG (H+L) Secondary Antibody, Rhodamine, Novus Biologicals™
Donkey Polyclonal Antibody
Brand: Novus Biologicals NBP1-73072
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Fluorophore-linked immunosorbent assay
Tekniske data
| IgG (H+L) | |
| Polyclonal | |
| Rhodamine | |
| Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free | |
| Mouse IgG whole molecule | |
| 2 mg | |
| Adaptive Immunity, Immunology | |
| Reconstitute with 1.0 mL deionized water (or equivalent). | |
| Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles. |
| Flow Cytometry, Immunocytochemistry/Immunofluorescence, Fluorophore-Linked Immunosorbent Assay | |
| LYOPH | |
| Flow Cytometry 1:500 - 1:2500, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Fluorophore-linked immunosorbent assay 1:10000 - 1:50000 | |
| Donkey | |
| Multi-step | |
| RUO | |
| Secondary | |
| Mouse | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion