missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Donkey anti-Human IgG (H+L) Secondary Antibody, Rhodamine (Pre-adsorbed), Novus Biologicals™
Donkey Polyclonal Antibody
Brand: Novus Biologicals NBP1-73616
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.
Tekniske data
| IgG (H+L) | |
| Polyclonal | |
| Rhodamine | |
| Donkey | |
| Affinity Purified | |
| RUO | |
| Secondary | |
| Reconstitute with 1.0 ml deionized water (or equivalent). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| LYOPH | |
| Flow Cytometry 1:1:10 - 1:1000, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Fluorophore-linked immunosorbent assay 1:10000 - 1:50000 | |
| Human IgG whole molecule | |
| 1 mg | |
| Adaptive Immunity, Immunology | |
| This antibody was pre-adsorbed against Bovine, Chicken, Goat, Guinea Pig, Hamster, Horse, Mouse, Rabbit, Rat, and Sheep Serum Proteins. No reaction was observed against Bovine, Chicken, Goat, Guinea Pig, Hamster, Horse, Mouse, Rabbit, Rat and Sheep Serum Proteins. | |
| Human | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion