Få mere at vide
Donepezil EP Impurity D, Mikromol™
Artikelnummer. 30052471
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30052471 100 mg
30051952 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30052471 Leverandør Mikromol Leverandørnr. MM0677.08
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Donepezil EP Impurity D
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5,6-Dimethoxy-2-[(4-pyridyl)methyl]indan-1-one
CAS 4803-57-0
Analyt- eller komponentnavne 5,6-Dimethoxy-2-[(4-pyridyl)methyl]indan-1-one
Forsendelsestilstand Room Temperature
Molekylær formel C17 H17 N O3
Certificeringer/compliance ISO 17025
Synonym 5,6-Dimethoxy-2-[(4-pyridyl)methyl]indan-1-one, 2,3-Dihydro-5,6-dimethoxy-2-(4-pyridinylmethyl)-1H-inden-1-one, 5,6-Dimethoxy-2-(4-pyridylmethyl)-1-indanone, Donepezil Hydrochloride Impurity D, Donepezil Impurity D
InChI formel InChI=1S/C17H17NO3/c1-20-15-9-12-8-13(7-11-3-5-18-6-4-11)17(19)14(12)10-16(15)21-2/h3-6,9-10,13H,7-8H2,1-2H3
SMIL COc1cc2CC(Cc3ccncc3)C(=O)c2cc1OC
IUPAC navn 5,6-dimethoxy-2-(pyridin-4-ylmethyl)-2,3-dihydroinden-1-one
Molekylvægt (g/mol) 283.32
Mængde 100 mg
Formel vægt 283.1208 g/mol