Få mere at vide
Domperidone, Thermo Scientific™
Peripheral dopamine receptor antagonist that does not cross the blood-brain barrier
Brand: Thermo Scientific Alfa Aesar J63681.MC
| Mængde | 100mg |
|---|
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsDomperidone is used as a Peripheral dopamine receptor antagonist that does not cross the blood-brain barrier. The antidopaminergic effects of Domperidone have been correlated to antiemetic and gastroprokinetic action.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Kemiske identifikatorer
| 57808-66-9 | |
| 425.917 | |
| FGXWKSZFVQUSTL-UHFFFAOYSA-N | |
| 3151 | |
| 6-chloro-3-[1-[3-(2-oxo-3H-benzimidazol-1-yl)propyl]piperidin-4-yl]-1H-benzimidazol-2-one |
| C22H24ClN5O2 | |
| MFCD00069256 | |
| domperidone, motilium, nauzelin, domperidonum, domperidona, domperidon, domperidonum inn-latin, domperidona inn-spanish, motillium, motinorm | |
| CHEBI:31515 | |
| C1CN(CCC1N2C3=C(C=C(C=C3)Cl)NC2=O)CCCN4C5=CC=CC=C5NC4=O |
Tekniske data
| Domperidone | |
| C22H24ClN5O2 | |
| 100mg | |
| domperidone, motilium, nauzelin, domperidonum, domperidona, domperidon, domperidonum inn-latin, domperidona inn-spanish, motillium, motinorm | |
| FGXWKSZFVQUSTL-UHFFFAOYSA-N | |
| 6-chloro-3-[1-[3-(2-oxo-3H-benzimidazol-1-yl)propyl]piperidin-4-yl]-1H-benzimidazol-2-one | |
| 3151 | |
| 425.91 |
| 57808-66-9 | |
| MFCD00069256 | |
| 14,3418 | |
| Slightly soluble in water. | |
| C1CN(CCC1N2C3=C(C=C(C=C3)Cl)NC2=O)CCCN4C5=CC=CC=C5NC4=O | |
| 425.917 | |
| CHEBI:31515 |
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.