missing translation for 'onlineSavingsMsg'
Få mere at vide

Domperidone, MedChemExpress

Artikelnummer. 16488286
Encompass_Preferred
Change view
Click to view available options
Mængde:
100 mg
200 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
16488286 100 mg
16431416 200 mg
16422126 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16488286 Supplier Medchem Express Supplier No. HYB0411

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Domperidone (R33812) is an orally active and selective dopamine-2 receptor antagonist. Domperidone acts as an antiemetic and a prokinetic agent through its effects on the chemoreceptor trigger zone and motor function of the stomach and small intestine.

Domperidone (R33812) is an orally active and selective dopamine-2 receptor antagonist. Domperidone acts as an antiemetic and a prokinetic agent through its effects on the chemoreceptor trigger zone and motor function of the stomach and small intestine.

Specifications

Kemisk navn eller materiale Domperidone
CAS 57808-66-9
Farve White
Renhedsgrad noter Research
Molekylær formel C22H24ClN5O2
Mængde 100 mg
Synonym R33812
Opløselighedsinformation DMSO : 50 mg/mL (117.40 mM; Need ultrasonic) ∣H2O : 0.67 mg/mL (1.57 mM; Need ultrasonic)
SMIL O=C1NC2=CC(Cl)=CC=C2N1C3CCN(CCCN4C5=CC=CC=C5NC4=O)CC3
Molekylvægt (g/mol) 425.91
Formel vægt 425.91
Procent renhed 99.79%
Grad Research
Fysisk form Solid
Til brug med (applikation) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.