Få mere at vide
Domperidone-d6, TRC
Artikelnummer. 30377582
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30377582 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30377582 Leverandør Toronto Research Chemicals Leverandørnr. TRCD53110210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Domperidone-d6
CAS 1329614-18-7
Molekylær formel C22H18D6ClN5O2
Mængde 10 mg
Synonym 5-Chloro-1-[1-[3-(2,3-dihydro-2-oxo-1H-benzimidazol-1-yl)propyl]-4-piperidinyl]-1,3-dihydro-2H-benzimidazol-2-one-d6, Euciton-d6, Evoxin-d6, Gastronorm-d6, KW 5338-d6, Mod-d6, Motilium-d6, NSC 299589-d6, Nauzelin-d6, Nauzelin Ds-d6, Peridon-d6, Peridys-d6, R 33812-d6
InChI formel InChI=1S/C22H24ClN5O2/c23-15-6-7-20-18(14-15)25-22(30)28(20)16-8-12-26(13-9-16)10-3-11-27-19-5-2-1-4-17(19)24-21(27)29/h1-2,4-7,14,16H,3,8-13H2,(H,24,29)(H,25,30)/i3D2,10D2,11D2
SMIL O=C1NC2=CC=CC=C2N1C([2H])([2H])C([2H])([2H])C([2H])([2H])N3CCC(N4C5=C(C=C(Cl)C=C5)NC4=O)CC3
Molekylvægt (g/mol) 431.95