Få mere at vide
Dofequidar fumarate, MedChemExpress
Artikelnummer. 16497226
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16497226 100 mg
16449787 5 mg
16447268 10 mg
16403287 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16497226 Leverandør Medchem Express Leverandørnr. HY17013A
MedChemExpress Dofequidar fumarate(MS-209 fumarate), an orally active quinoline compound, has been reported to overcome MDR by inhibiting ABCB1/P-gp, ABCC1/MDR-associated protein 1, or both.
Dofequidar fumarate(MS-209 fumarate), an orally active quinoline compound, has been reported to overcome MDR by inhibiting ABCB1/P-gp, ABCC1/MDR-associated protein 1, or both.
Tekniske data
Kemisk navn eller materiale Dofequidar fumarate
CAS 153653-30-6
Farve White
Renhedsgrad noter Research
Molekylær formel C34H35N3O7
Mængde 100 mg
Synonym MS-209
Opløselighedsinformation DMSO : 100 mg/mL (167.32 mM; Need ultrasonic) ∣H2O : 1 mg/mL (1.67 mM; Need ultrasonic)
SMIL OC(COC1=C2C=CC=NC2=CC=C1)CN3CCN(C(C(C4=CC=CC=C4)C5=CC=CC=C5)=O)CC3.O=C(O)/C=C/C(O)=O
Molekylvægt (g/mol) 597.66
Formel vægt 597.66
Procent renhed 98.4%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-programmed cell death