Få mere at vide
Dodecyltrimethylammonium-d3 Chloride, TRC
Artikelnummer. 30370024
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30370024 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30370024 Leverandør Toronto Research Chemicals Leverandørnr. TRCD52550210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Dodecyltrimethylammonium-d3 Chloride
CAS 1219337-20-8
Molekylær formel C15H31D3ClN
Mængde 10 mg
Synonym Dodecyltrimethylammonium-d3 chloride, Dodecyldimethyl(methyl-d3)ammonium chloride, Lauryldimethyl(methyl-d3)ammonium chloride, Dodecyltrimethylammonium chloride-d3, Dodecyltrimethylammonium-D3 (N-methyl-D3) chloride, N,N-Dimethyl-N-(methyl-d3)-1-dodecanaminium chloride
InChI formel InChI=1S/C15H34N.ClH/c1-5-6-7-8-9-10-11-12-13-14-15-16(2,3)4;/h5-15H2,1-4H3;1H/q+1;/p-1/i2D3;
SMIL [Cl-].[2H]C([2H])([2H])[N+](C)(C)CCCCCCCCCCCC
Molekylvægt (g/mol) 266.91