Få mere at vide
Dodecylbenzenesulfonic Acid (70 wt. % in isopropanol, Technical grade, Mixture of different chain lengths), TRC
Artikelnummer. 30136969
Skift visning
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136969 1 g
30010173 500 mg
30012290 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136969 Leverandør Toronto Research Chemicals Leverandørnr. TRCD5256631G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 27176-87-0
Molekylær formel C18H30O3S
Molekylvægt (g/mol) 326.5
Synonym Dodecyl-Benzenesulfonic acid, 2-4248S, A 40S, ABS 100, Ambicat LE 4476, B 121, B 121 (surfactant), Bio-Soft LA acid, Bio-Soft S 100, Bio-Soft S 101, Bio-Soft S 126, CATA CFC 5019KC, Calsoft LAS 99, Cat 6000, Catalyst 600, Catalyst 6000, Cycat 600, DBS, Dobanic acid, Dow Corning 2-4248S, E 7256, Elfan WA Sulfonic Acid, LABS, LABS (detergent), LAS 99, LE 4476, Laurylbenzenesulfonic acid, Lipon LH 500, Maranil DBS, Marlon AS 3, Nacconol 98SA, Nacure 5074, Nacure 5076, Nansa 1042, Nansa 1042P, Nansa SSA, Neopelex FS, Neopelex GS, Neopelex GSP, P 3 Vetralat, Pelex F 25, Polystep A 13, Rhodacal SSA/A, Richonic Acid B, S 100, Soft Osen 5S, Statsafe 6000, Sulfosoft, Sulframin 1298, Sulframin Acid 1298, Taycacure AC 430, Taycapower L 120D, Taycapower L 121, Taycapower L 122, Taycatox 120, Ufacid K, Vanwet acid 98, Witco 1298, Witco 1298 Acid Soft, Witco 1298 Soft Acid, Witconic 1298 Hard Acid, Witconic 1298 Soft Acid, Witconic 1298H, n-Dodecylbenzenesulfonic acid
SMIL CCCCCCCCCCCCc1ccccc1S(=O)(=O)O
Tekniske data
Kemisk navn eller materiale Dodecylbenzenesulfonic Acid (Technical grade)
Mængde 1 g
InChI formel InChI=1S/C18H30O3S/c1-3-5-6-7-8-9-10-11-16(4-2)17-12-14-18(15-13-17)22(19,20)21/h12-16H,3-11H2,1-2H3,(H,19,20,21)
Koncentration eller sammensætning (efter analyt eller komponenter) Dodecylbenzenesulfonic Acid (Technical grade) 70%