Få mere at vide
Dodecanedioic-d20 Acid, TRC
Artikelnummer. 30371315
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30371315 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30371315 Leverandør Toronto Research Chemicals Leverandørnr. TRCD49453350MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Dodecanedioic-d20 Acid
CAS 89613-32-1
Molekylær formel C12 D20 H2 O4
Mængde 50 mg
Synonym 1,10-Decanedicarboxylic Acid-d20, 1,10-Dicarboxydecane-d20, 1,12-Dodecanedioic Acid-d20, Corfree M 2-d20, Decamethylenedicarboxylic Acid-d20, NSC 400242-d20, SL-AH-d20, n-Dodecane-α,ω-dioic Acid-d20, n-Dodecanedioic Acid-d20
InChI formel InChI=1S/C12H22O4/c13-11(14)9-7-5-3-1-2-4-6-8-10-12(15)16/h1-10H2,(H,13,14)(H,15,16)/i1D2,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2
SMIL [2H]C([2H])(C(=O)O)C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C(=O)O
IUPAC navn 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11-icosadeuteriododecanedioic acid
Molekylvægt (g/mol) 250.42
Formel vægt 250.2773