missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Dodecanedioic Acid 1-Methyl Ester, TRC
Specifications
Specifications
| Kemisk navn eller materiale | Dodecanedioic Acid 1-Methyl Ester |
| CAS | 3903-40-0 |
| Molekylær formel | C13 H24 O4 |
| Mængde | 500 mg |
| Synonym | 11-Carbomethoxyundecanoic Acid, Methyl 11-Carboxyundecanoate, Monomethyl 1,12-Dodecanedioate, Monomethyl Dodecanedioate, NSC 29000 |
| InChI formel | InChI=1S/C13H24O4/c1-17-13(16)11-9-7-5-3-2-4-6-8-10-12(14)15/h2-11H2,1H3,(H,14,15) |
| SMIL | COC(=O)CCCCCCCCCCC(=O)O |
| IUPAC navn | 12-methoxy-12-oxododecanoic acid |
| Molekylvægt (g/mol) | 244.33 |
| Formel vægt | 244.1675 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?