Få mere at vide
Dodecane-d26, TRC
Artikelnummer. 30381643
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30381643 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30381643 Leverandør Toronto Research Chemicals Leverandørnr. TRCD494627100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale n-Dodecane-d26
CAS 16416-30-1
Molekylær formel C12 D26
Mængde 100 mg
Synonym Dodecane-1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-d26, Perdeuterododecane, Adakane 12-d26, Ba 51-090453-d26, C12-n-Alkane-d26, Cactus Normal Paraffin N 12D-d26, NSC 8714-d26, Parafol 12-97-d26, Parafol C12-97-d26, Dodecane-d26 (8CI,9CI), Dodecane-d26, Dodecane D26
InChI formel InChI=1S/C12H26/c1-3-5-7-9-11-12-10-8-6-4-2/h3-12H2,1-2H3/i1D3,2D3,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H]
IUPAC navn 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-hexacosadeuteriododecane
Molekylvægt (g/mol) 196.5
Formel vægt 196.3666