Få mere at vide
Docusate Potassium, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Docusate Potassium |
| CAS | 7491-09-0 |
| Molekylær formel | C20 H37 O7 S . K |
| Mængde | 100 mg |
| Synonym | Docusate Potassium, Sulfobutanedioic Acid 1,4-Bis(2-ethylhexyl) Ester Potassium Salt, Sulfosuccinic Acid 1,4-Bis(2-ethylhexyl) Ester Potassium Salt, Bis(2-ethylhexyl) Potassium Sulfosuccinate, Bis-2-ethylhexyl-2-sulfobutane-1,4-dioate Potassium Salt, Dioctyl Potassium Sulfosuccinate, Docusate Potassium, Potassium Bis(2-ethylhexyl) Sulfosuccinate, Potassium di(2-Ethylhexyl) Sulfosuccinate, Potassium Dioctyl Sulfosuccinate, Rectalad Enema |
| InChI formel | InChI=1S/C20H38O7S.K/c1-5-9-11-16(7-3)14-26-19(21)13-18(28(23,24)25)20(22)27-15-17(8-4)12-10-6-2;/h16-18H,5-15H2,1-4H3,(H,23,24,25);/q;+1/p-1 |
| SMIL | [K+].CCCCC(CC)COC(=O)CC(C(=O)OCC(CC)CCCC)S(=O)(=O)[O-] |
| IUPAC navn | potassium;1,4-bis(2-ethylhexoxy)-1,4-dioxobutane-2-sulfonate |
| Molekylvægt (g/mol) | 460.67 |
| Formel vægt | 460.1897 |