Få mere at vide
Docosanoic-7,7,8,8-d4 Acid, TRC
Artikelnummer. 30330283
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30330283 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30330283 Leverandør Toronto Research Chemicals Leverandørnr. TRCB1195911MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Docosanoic-7,7,8,8-d4 Acid
CAS 1193721-65-1
Molekylær formel C22 D4 H40 O2
Mængde 1 mg
Synonym Docosanoic-7,7,8,8-d4 acid (ACI)
InChI formel InChI=1S/C22H44O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22(23)24/h2-21H2,1H3,(H,23,24)/i15D2,16D2
SMIL [2H]C([2H])(CCCCCCCCCCCCCC)C([2H])([2H])CCCCCC(=O)O
IUPAC navn 7,7,8,8-tetradeuteriodocosanoic acid
Molekylvægt (g/mol) 344.6082
Formel vægt 344.3592