Få mere at vide
Docosanoic-7,7,8,8-d4 Acid, CDN
Artikelnummer. 30315989
Skift visning
Mængde:
0.05 g
0.1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30315989 0.1 g
30312293 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30315989 Leverandør CDN Leverandørnr. CDND57090.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Docosanoic-7,7,8,8-d4 Acid
CAS 1193721-65-1
Molekylær formel C22 D4 H40 O2
Mængde 0.1 g
Synonym Docosanoic-7,7,8,8-d4 acid (ACI)
InChI formel InChI=1 S/C22H44O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22(23)24/h2-21H2,1H3,(H,23,24)/i15D2,16D2
SMIL [2 H]C([2 H])(CCCCCCCCCCCCCC)C([2 H])([2 H])CCCCCC(=O)O
IUPAC navn 7,7,8,8-tetradeuteriodocosanoic acid
Molekylvægt (g/mol) 344.6082
Formel vægt 344.3592 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity