missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Docosane, Sigma-Aldrich™
Tekniske data
Tekniske data
| CAS | 629-97-0 |
| Smeltepunkt | 44°C |
| Tæthed | 0.794 g/cm3 at 20°C |
| Kogepunkt | 369°C/1013 hPa |
| Molekylær formel | CH3(CH2)20CH3 |
| MDL nummer | MFCD00009348 |
| Mængde | 50 g |
| Synonym | Docosane, Docosane |
| Molekylvægt (g/mol) | 310.6 |
| Grad | Research |
Produkttitel
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?