Få mere at vide
Docosahexaenoic Acid N-Succinimide, TRC
Artikelnummer. 30373832
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30373832 250 mg
30373164 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30373832 Leverandør Toronto Research Chemicals Leverandørnr. TRCD494595250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 160801-26-3
Molekylær formel C26H35NO4
Molekylvægt (g/mol) 425.56
Synonym (all-Z)-1-[(1-Oxo-4,7,10,13,16,19-docosahexaenyl)oxy]-2,5-pyrrolidinedione
IUPAC navn 2,5-dioxopyrrolidin-1-yl (4Z,7Z,10Z,13Z,16Z,19Z)-docosa-4,7,10,13,16,19-hexaenoate
SMIL O(C(CC/C=C\C/C=C\C/C=C\C/C=C\C/C=C\C/C=C\CC)=O)N1C(=O)CCC1=O
Tekniske data
Kemisk navn eller materiale Docosahexaenoic Acid N-Succinimide
Mængde 250 mg
InChI formel InChI=1S/C26H35NO4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-26(30)31-27-24(28)22-23-25(27)29/h3-4,6-7,9-10,12-13,15-16,18-19H,2,5,8,11,14,17,20-23H2,1H3/b4-3-,7-6-,10-9-,13-12-,16-15-,19-18-
Formel vægt 425.26