Få mere at vide
Docosahexaenoic Acid-d5 (>80%), TRC
Artikelnummer. 30378738
Skift visning
Mængde:
2.5 mg
Artikelnummer. Mængde
30378738 2.5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30378738 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4944992.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Docosahexaenoic Acid-d5
CAS 1197205-71-2
Molekylær formel C22 2H5 H27 O2
Mængde 2.5 mg
Synonym 4,7,10,13,16,19-Docosahexaenoic-21,21,22,22,22-d5 acid, (4Z,7Z,10Z,13Z,16Z,19Z)-, (4Z,7Z,10Z,13Z,16Z,19Z)-4,7,10,13,16,19-Docosahexaenoic-21,21,22,22,22-d5 acid, (all-Z)-4,7,10,13,16,19-Docosahexaenoic-21,21,22,22,22-d5 acid, Docosahexaenoic acid-d5, all-cis-4,7,10,13,16,19-Docosahexaenoic-d5 acid, cis-4,7,10,13,16,19-Docosahexaenoic Acid D5 (ethyl-D5)
InChI formel InChI=1S/C22H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22(23)24/h3-4,6-7,9-10,12-13,15-16,18-19H,2,5,8,11,14,17,20-21H2,1H3,(H,23,24)/b4-3-,7-6-,10-9-,13-12-,16-15-,19-18-/i1D3,2D2
SMIL [2H]C([2H])([2H])C([2H])([2H])\C=C/C\C=C/C\C=C/C\C=C/C\C=C/C\C=C/CCC(=O)O
IUPAC navn (4Z,7Z,10Z,13Z,16Z,19Z)-21,21,22,22,22-pentadeuteriodocosa-4,7,10,13,16,19-hexaenoic acid
Molekylvægt (g/mol) 333.52
Formel vægt 333.2716