Få mere at vide
Docetaxel Hydrate, TRC
Artikelnummer. 30009580
Skift visning
Mængde:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30009580 250 mg
30007421 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30009580 Leverandør Toronto Research Chemicals Leverandørnr. TRCD494420250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 700367-34-6
Molekylær formel C43 H53 N O14 . H2 O
Molekylvægt (g/mol) 825.89
Synonym Benzenepropanoic acid, β-[[(1,1-dimethylethoxy)carbonyl]amino]-α-hydroxy-, (2aR,4S,4aS,6R,9S,11S,12S,12aR,12bS)-12b-(acetyloxy)-12-(benzoyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-4,6,11-trihydroxy-4a,8,13,13-tetramethyl-5-oxo-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl ester, hydrate, (αR,βS)- (9CI), Docetaxel hydrate
SMIL O.CC(=O)O[C@@]12CO[C@@H]1C[C@H](O)[C@]3(C)[C@@H]2[C@H](OC(=O)c4ccccc4)[C@]5(O)C[C@H](OC(=O)[C@H](O)[C@@H](NC(=O)OC(C)(C)C)c6ccccc6)C(=C([C@@H](O)C3=O)C5(C)C)C
Tekniske data
Kemisk navn eller materiale Docetaxel Hydrate
Mængde 250 mg
InChI formel InChI=1S/C43H53NO14.H2O/c1-22-26(55-37(51)32(48)30(24-15-11-9-12-16-24)44-38(52)58-39(3,4)5)20-43(53)35(56-36(50)25-17-13-10-14-18-25)33-41(8,34(49)31(47)29(22)40(43,6)7)27(46)19-28-42(33,21-54-28)57-23(2)45;/h9-18,26-28,30-33,35,46-48,53H,19-21H2,1-8H3,(H,44,52);1H2/t26-,27-,28+,30-,31+,32+,33-,35-,41+,42-,43+;/m0./s1
Formel vægt 825.3572