Få mere at vide
Docetaxel-d9, TRC
Artikelnummer. 30377630
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30377630 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30377630 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4944221MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale d9-Docetaxel
CAS 940867-25-4
Renhedsgrad noter HPLC
Molekylær formel C43 D9 H44 N O14
Mængde 1 mg
Synonym Docetaxel-d9, Docetaxel D9 (tert-butyl D9)
InChI formel InChI=1S/C43H53NO14/c1-22-26(55-37(51)32(48)30(24-15-11-9-12-16-24)44-38(52)58-39(3,4)5)20-43(53)35(56-36(50)25-17-13-10-14-18-25)33-41(8,34(49)31(47)29(22)40(43,6)7)27(46)19-28-42(33,21-54-28)57-23(2)45/h9-18,26-28,30-33,35,46-48,53H,19-21H2,1-8H3,(H,44,52)/t26-,27-,28+,30-,31+,32+,33-,35-,41+,42-,43+/m0/s1/i3D3,4D3,5D3
SMIL [2H]C([2H])([2H])C(OC(=O)N[C@H]([C@@H](O)C(=O)O[C@H]1C[C@@]2(O)[C@@H](OC(=O)c3ccccc3)[C@@H]4[C@@]5(CO[C@@H]5C[C@H](O)[C@@]4(C)C(=O)[C@H](O)C(=C1C)C2(C)C)OC(=O)C)c6ccccc6)(C([2H])([2H])[2H])C([2H])([2H])[2H]
Molekylvægt (g/mol) 816.93
Formel vægt 816.4031
Procent renhed >95