Få mere at vide
DNCIC1 Antibody [PerCP], Novus Biologicals Biologicals™
Artikelnummer. 30498743 Shop alle Bio Techne produkter
Skift visning
Mængde:
0.1 mL
Artikelnummer. Mængde
30498743 0.1 mL
1 options
Artikelnummer. 30498743 Leverandør Novus Biologicals Leverandørnr. NBP338053PCP
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Rabbit Polyclonal Antibody
DNCIC1 Polyclonal antibody specifically detects DNCIC1 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Tekniske data
Antigen DNCIC1
Applikationer ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Klassifikation Polyclonal
Konjugeret PerCP
Formulering PBS
Gene Alias cytoplasmic dynein 1 intermediate chain 1, Cytoplasmic dynein intermediate chain 1, DNCI1cytoplasmic, intermediate polypeptide 1, DNCIC1DH IC-1, Dynein intermediate chain 1, cytosolic, dynein, cytoplasmic 1, intermediate chain 1
Værtsarter Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human DNCIC1 (NP_001129028.1).,, Sequence:, MSDKSDLKAELERKKQRLAQIREEKKRKEEERKKKEADMQQKKEPVQDDSDLDRKRRETEALLQSIGISPEPPLVPTPMSPSSKSVSTPSEAGSQDSGDLGPLTRTLQWDTDPSVLQLQSDSELGRRLHKLGVSKVTQVDFLPREVVSYSKETQTPLATHQSEEDEEDEEMVESKVGQDSELENQDKKQEVKEAPPRELTEEEKQQILHSEEFLIFFDRT
Oprensningsmetode Affinity purified
Mængde 0.1 mL
Regulatorisk status RUO
Forskningsdisciplin Cell Biology, Cellular Markers, Golgi Apparatus Markers, Neuronal Cell Markers
Primær eller sekundær Primary
Gen-id (Entrez) 1780
Målarter Human, Mouse, Rat
Indhold og opbevaring Store at 4°C in the dark.
Produkttype Antibody
Form Purified
Isotype IgG