missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJB12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-87285
Additional Details : Weight : 0.00970kg
Description
DNAJB12 Polyclonal specifically detects DNAJB12 in Mouse samples. It is validated for Western Blot.Specifications
DNAJB12 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
54788 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
DJ10FLJ20027, DKFZp586B2023, DnaJ (Hsp40) homolog, subfamily B, member 12, dnaJ homolog subfamily B member 12, FLJ0027 | |
The immunogen is a synthetic peptide directed towards the N-terminal region of Mouse DNAJB12 | |
100 μL | |
Primary | |
Mouse | |
Purified |