missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
DNA Polymerase delta, catalytic subunit Antibody (607), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-476
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
DNA Polymerase delta, catalytic subunit Monoclonal specifically detects DNA Polymerase delta, catalytic subunit in Human, Mouse samples. It is validated for Western Blot.
Tekniske data
| DNA Polymerase delta, catalytic subunit | |
| Monoclonal | |
| 1.0mg/mL | |
| Western Blot 1:1000 | |
| POLD1 | |
| Recombinant protein encoding amino acids 1-140 of the large subunit of human DNA polymerase d. | |
| 100 μL | |
| Breast Cancer, Core ESC Like Genes, DNA Polymerases, DNA Repair, Stem Cell Markers | |
| 5424 | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| 607 | |
| Unconjugated | |
| CDC2, CDC2 homolog, DNA polymerase delta catalytic subunit, DNA polymerase subunit delta p125, DNA-directed polymerase delta 1, EC 2.7.7.7, POLD, polymerase (DNA directed), delta 1, catalytic subunit (125kD), polymerase (DNA directed), delta 1, catalytic subunit 125kDa | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| DNA Polymerase delta (607). Cross-reacts with Human and Mouse. Not yet tested in other species. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion