Learn More
Lonza™ DNA Markers
For rapid size estimation of PCR products and restriction fragments
Brand: Lonza™ 50631
Additional Details : Weight : 2.94830kg
Specifications
130 ng/μL | |
Accurate size estimation of DNA fragments | |
13 | |
4°C |
DNA Molecular Weight Marker/Ladder | |
2 x 250μL DNA Marker, 250μL 6X loading Buffer contains Bromophenol Blue, 1.1mL Triple Dye Loading Buffer 6X contains Orange G, Bromophenol Blue, Xylene Cyanol | |
250 μL |