Learn More
Lonza™ DNA Markers
For rapid size estimation of PCR products and restriction fragments
Brand:  Lonza™ 50631
Additional Details : Weight : 2.94830kg
 View more versions of this product
Product Code. 11587745
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
Specifications
Specifications
130 ng/μL
Accurate size estimation of DNA fragments
13
4°C
DNA Molecular Weight Marker/Ladder
2 x 250μL DNA Marker, 250μL 6X loading Buffer contains Bromophenol Blue, 1.1mL Triple Dye Loading Buffer 6X contains Orange G, Bromophenol Blue, Xylene Cyanol
250 μL