missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ DNA ligase IV Polyclonal Antibody
Beskrivelse
Recommended positive controls: U87-MG, mouse brain, rat brain. Predicted reactivity: Mouse (82%), Rat (81%), Sheep (83%), Bovine (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a DNA ligase that joins single-strand breaks in a double-stranded polydeoxynucleotide in an ATP-dependent reaction. This protein is essential for V(D)J recombination and DNA double-strand break (DSB) repair through nonhomologous end joining (NHEJ). This protein forms a complex with the X-ray repair cross complementing protein 4 (XRCC4), and further interacts with the DNA-dependent protein kinase (DNA-PK). Both XRCC4 and DNA-PK are known to be required for NHEJ. The crystal structure of the complex formed by this protein and XRCC4 has been resolved. Defects in this gene are the cause of LIG4 syndrome. Alternatively spliced transcript variants encoding the same protein have been observed.
Tekniske data
Tekniske data
| Antigen | DNA ligase IV |
| Applikationer | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Klassifikation | Polyclonal |
| Koncentration | 1 mg/mL |
| Konjugeret | Unconjugated |
| Formulering | 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | LIG4 |
| Genadgangsnr. | P49917, Q8BTF7 |
| Gene Alias | 5830471N16Rik; DNA joinase; DNA ligase (ATP) 4; DNA ligase 4; DNA ligase IV; DNA repair enzyme; Lig4; LIG4S; ligase IV, DNA, ATP-dependent; polydeoxyribonucleotide synthase [ATP] 4; polynucleotide ligase; sealase; tiny |
| Gensymboler | LIG4 |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?