Learn More
Cytiva (formerly Pall Lab) DMSO-Safe Acrodisc™ Syringe Filters
Polypropylene housing and nylon membrane are compatible with solutions containing DMSO.
Brand: Cytiva (formerly Pall Lab) 4433
1320.50 DKK valid until 2024-06-30
Use promo code "21742" to get your promotional price.
Additional Details : Weight : 0.56050kg
Description
- Chemical compatibility with esters, bases, and alcohols
- Not approved for use in medical, clinical, surgical or other patient applications
Specifications
Nylon | |
Sterile | |
Polypropylene |
0.2 μm | |
2.8 cm2 | |
25 mm |