Få mere at vide
Tocris Bioscience™ DMOG
Artikelnummer. 18025342
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18025342 10 mg
18064892 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18025342 Leverandør Tocris Bioscience™ Leverandørnr. 4408/10
Prolyl hydroxylase inhibitor
DMOG is a prolyl 4-hydroxylase (P4H) inhibitor; inhibits hypoxia-inducible factor α (HIF-α) prolyl hydroxylase (HIF-PH). Increases levels of HIF-1α; promotes cell survival under hypoxic conditions. Inhibits differentiation of hematopoietic stem cells (HSCs) in culture.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 89464-63-1
Molekylær formel C6H9NO5
Tekniske data
Kemisk navn eller materiale DMOG
Mængde 10 mg
Formel vægt 175.14
Renhed 0.97
Molekylvægt (g/mol) 175.14