missing translation for 'onlineSavingsMsg'
Få mere at vide

DMOG, MedChemExpress

Artikelnummer. 16481786
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
100 mg
200 mg
500 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16481786 500 mg
16452967 50 mg
16451206 200 mg
16426986 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16481786 Leverandør Medchem Express Leverandørnr. HY15893

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress DMOG (Dimethyloxallyl Glycine) is a cell permeable and competitive inhibitor of HIF-PH, which results in HIF-1α stabilisation and accmulation in vitro and in vivo. DMOG is an α-ketoglutarate analogue and inhibits α-KG-dependent hydroxylases. DMOG acts as a pro-angiogenic agent and plays a protective role in experimental model of colitis and diarrhoea via HIF-1 related signal. DMOG induces cell autophagy.

DMOG (Dimethyloxallyl Glycine) is a cell permeable and competitive inhibitor of HIF-PH, which results in HIF-1α stabilisation and accmulation in vitro and in vivo. DMOG is an α-ketoglutarate analogue and inhibits α-KG-dependent hydroxylases. DMOG acts as a pro-angiogenic agent and plays a protective role in experimental model of colitis and diarrhoea via HIF-1 related signal. DMOG induces cell autophagy.

Tekniske data

Kemisk navn eller materiale DMOG
CAS 89464-63-1
Farve White
Renhedsgrad noter Research
Molekylær formel C6H9NO5
Mængde 500 mg
Synonym Dimethyloxallyl Glycine
Opløselighedsinformation DMSO : ≥ 50 mg/mL (285.49 mM) ∣H2O : 50 mg/mL (285.49 mM; Need ultrasonic)
SMIL O=C(OC)CNC(C(OC)=O)=O
Molekylvægt (g/mol) 175.14
Formel vægt 175.14
Procent renhed 98.0%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-programmed cell death
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.