Få mere at vide
Tocris Bioscience™ DMHBO+
Beskrivelse
DMHBO+ is a cationic fluorophore. Its fluorescence is activated by binding to chili aptamer (Kd = 12 nM). Chili-DMHBO+ complex mimics red fluorescent proteins, suitable for imaging RNA in cells. It is an ideal FRET donor to the rhodamine dye Atto 590 and has applications in FRET-based analytical RNA systems. Excitation/emission maxima (λ) = 456/592 nm; quantum yield = 0.1; stokes shift = 136 nm.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 4-(4-((Z)-4-Hydroxy-3,5-dimethoxybenzylidene)-2-((hydroxyimino)methyl)-5-oxo-4,5-dihydro-1H-imidazol-1-yl)-N,N,N-trimethylbenzenaminium iodide |
| CAS | 2322286-81-5 |
| Mængde | 10 mg |
| Mål | Light-up Aptamers |
| Molekylær formel | C22H25IN4O5 |
| Renhed | 98% |