missing translation for 'onlineSavingsMsg'
Få mere at vide

R&DSystems™ DMEM, low glucose, 25 mM HEPES, L-glutamine, no sodium pyruvate

Artikelnummer. 18471600
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
500 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
18471600 500 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18471600 Supplier R&DSystems™ Supplier No. M17050

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

This item is not returnable. View return policy

Dulbecco’s Modified Eagle Medium (DMEM) with 1000 mg/L D-glucose, 25 mM HEPES buffer, and L-glutamine. Without sodium pyruvate.

TRUSTED_SUSTAINABILITY

Specifications

Til brug med (applikation) Cell Culture
Mængde 500 ml
Renhed eller kvalitetsklasse 0.2 μm filtered
Indhold og opbevaring 2 - 8 °C, protected from light
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.